Ethyl 2-(pyrazin-2-yl)acetate 95%
Leverantör: Apollo Scientific
OR953240-1G OR953240-5G
APOSOR953240-1GEA 2250 SEK
APOSOR953240-1G APOSOR953240-5G
Ethyl 2-(pyrazin-2-yl)acetate 95%
Ethyl 2-(pyrazin-2-yl)acetate
Formel: C₈H₁₀N₂O₂
MW: 166.177 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10698624
CAS nummer: 1060815-23-7