1,2,3,4-Tetrahydro-1,5-naphthyridine 97%
Leverantör: Apollo Scientific
OR953110-1G OR953110-250MG
APOSOR953110-1GEA 1300 SEK
APOSOR953110-1G APOSOR953110-250MG
1,2,3,4-Tetrahydro-1,5-naphthyridine 97%
1,2,3,4-Tetrahydro-1,5-naphthyridine
Formel: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08444439
CAS nummer: 13993-61-8