Ethyl 4-methylpyrimidine-5-carboxylate 96%
Leverantör: Apollo Scientific
OR952976-1G OR952976-5G
APOSOR952976-1GEA 705 SEK
APOSOR952976-1G APOSOR952976-5G
Ethyl 4-methylpyrimidine-5-carboxylate 96%
Ethyl 4-methylpyrimidine-5-carboxylate
Formel: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07369107
CAS nummer: 110960-73-1