3-Ethoxy-2-hydroxy-benzoic acid 95%

Leverantör: Apollo Scientific

Synonyms: 3-Ethoxy-2-hydroxybenzoic acid

OR952884-1G OR952884-250MG
APOSOR952884-1GEA 7625 SEK
APOSOR952884-1G APOSOR952884-250MG
3-Ethoxy-2-hydroxy-benzoic acid 95%
3-Ethoxy-2-hydroxy-benzoic acid
Formel: C₉H₁₀O₄
MW: 182,173 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01412200
CAS nummer: 67127-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR