5-Amino-1-isopropyl-1H-indazole 95%
Leverantör: Apollo Scientific
OR951572-1G OR951572-250MG
APOSOR951572-1GEA 5900 SEK
APOSOR951572-1G APOSOR951572-250MG
5-Amino-1-isopropyl-1H-indazole 95%
5-Amino-1-isopropyl-1H-indazole
Formel: C₁₀H₁₃N₃
MW: 175,23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09026344
CAS nummer: 928821-18-5