(S)-1-o-Tolylethanamine hydrochloride, tech
Leverantör: Apollo Scientific
Varning
Synonyms: (S)-1-(o-Tolyl)ethanamine hydrochloride
OR951537-100MG OR951537-250MG
APOSOR951537-100MGEA 820 SEK
APOSOR951537-100MG APOSOR951537-250MG
(S)-1-o-Tolylethanamine hydrochloride, tech
(S)-1-o-Tolylethanamine hydrochloride
Formel: C₉H₁₃N·ClH
MW: 171.667 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11101268
CAS nummer: 1332832-16-2