Ethyl 1-methylimidazole-5-carboxylate 98%
Leverantör: Apollo Scientific
Synonyms: Ethyl 1-Methylimidazole-5-carboxylate
OR951535-1G OR951535-5G
APOSOR951535-1GEA 1975 SEK
APOSOR951535-1G APOSOR951535-5G
Ethyl 1-methylimidazole-5-carboxylate 98%
Ethyl 1-methylimidazole-5-carboxylate
Formel: C₇H₁₀N₂O₂
MW: 154,166 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06204805
CAS nummer: 66787-70-0