6-Hydroxy-5-methoxynicotinic acid 96%
Leverantör: Apollo Scientific
OR951302-100MG OR951302-1G OR951302-250MG
APOSOR951302-100MGEA 2075 SEK
APOSOR951302-100MG APOSOR951302-1G APOSOR951302-250MG
6-Hydroxy-5-methoxynicotinic acid 96%
6-Hydroxy-5-methoxynicotinic acid
Formel: C₇H₇NO₄
MW: 169,135 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12401649
CAS nummer: 1171919-98-4