4-N-Alloc-aminocyclohexanone 95%
Leverantör: Apollo Scientific
Synonyms: Allyl (4-oxocyclohexyl)carbamate
OR951103-1G OR951103-5G
APOSOR951103-1GEA 685 SEK
APOSOR951103-1G APOSOR951103-5G
4-N-Alloc-aminocyclohexanone 95%
4-N-Alloc-aminocyclohexanone
Formel: C₁₀H₁₅NO₃
MW: 197.231 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02183577
CAS nummer: 391248-19-4