3-Methylisonicotinamide 95%
Leverantör: Apollo Scientific
OR950771-1G
OR950771-250MG
APOSOR950771-1GEA
2450
SEK
APOSOR950771-1G
APOSOR950771-250MG
3-Methylisonicotinamide 95%
3-Methylisonicotinamide
Formel:
C₇H₈N₂O MW: 136,151 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD03411690 CAS nummer: 251101-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...