cis-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
Leverantör: Apollo Scientific
Synonyms: cis-Methyl 3-hydroxy-1-methylcyclobutanecarboxylate
OR949331-1G OR949331-250MG OR949331-500MG
APOSOR949331-1GEA 14075 SEK
APOSOR949331-1G APOSOR949331-250MG APOSOR949331-500MG
cis-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
cis-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
Formel: C₇H₁₂O₃
MW: 144,168 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23105987
CAS nummer: 626238-08-2