1,2-Dihydro-1-(4-methoxyphenyl)-2-oxopyridine-3-carbonitrile 96%
Leverantör: Apollo Scientific
Synonyms: 3-Cyano-1,2-dihydro-1-(4-methoxyphenyl)-2-oxopyridine, 4-(3-Cyano-2-oxopyridin-1(2H)-yl)anisole
OR9491-1G OR9491-250MG
APOSOR9491-1GEA 6000 SEK
APOSOR9491-1G APOSOR9491-250MG
1,2-Dihydro-1-(4-methoxyphenyl)-2-oxopyridine-3-carbonitrile 96%
1,2-Dihydro-1-(4-methoxyphenyl)-2-oxopyridine-3-carbonitrile
Formel: C₁₃H₁₀N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD09258785
CAS nummer: 929000-87-3