4,6-Dimethyl-2,3-dihydropyridazin-3-one
Leverantör: Apollo Scientific
OR948323-1G OR948323-250MG OR948323-500MG
APOSOR948323-1GEA 10625 SEK
APOSOR948323-1G APOSOR948323-250MG APOSOR948323-500MG
4,6-Dimethyl-2,3-dihydropyridazin-3-one
4,6-Dimethyl-2,3-dihydropyridazin-3-one
Formel: C₆H₈N₂O
MW: 124,141 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00867544
CAS nummer: 7007-92-3