Ethyl 4-methyl-2-cyclohexanone-1-carboxylate 95%
Leverantör: Apollo Scientific
OR946697-1G OR946697-5G
APOSOR946697-1GEA 1975 SEK
APOSOR946697-1G APOSOR946697-5G
Ethyl 4-methyl-2-cyclohexanone-1-carboxylate 95%
Ethyl 4-methyl-2-cyclohexanone-1-carboxylate
Formel: C₁₀H₁₆O₃
Storage Temperature: Omgivande
MDL Number: MFCD00001632
CAS nummer: 13537-82-1