2-Chloromethyl-piperidine hydrochloride 95%
Leverantör: Apollo Scientific
OR946549-1G OR946549-250MG
APOSOR946549-1GEA 9075 SEK
APOSOR946549-1G APOSOR946549-250MG
2-Chloromethyl-piperidine hydrochloride 95%
2-Chloromethyl-piperidine hydrochloride
Formel: C₆H₁₂ClN·ClH
MW: 170,08 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02667639
CAS nummer: 2554-93-0