4-Methoxy-4-methyl-piperidine hydrochloride 97%
Leverantör: Apollo Scientific
Synonyms: 4-Methoxy-4-methylpiperidine hydrochloride
OR944010-1G OR944010-250MG OR944010-500MG
APOSOR944010-1GEA 8475 SEK
APOSOR944010-1G APOSOR944010-250MG APOSOR944010-500MG
4-Methoxy-4-methyl-piperidine hydrochloride 97%
4-Methoxy-4-methyl-piperidine hydrochloride
Formel: C₇H₁₅NO·ClH
MW: 165,661 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12755739
CAS nummer: 3970-73-8