5,6-Dimethylnicotinonitrile 97%
Leverantör: Apollo Scientific
Varning
OR943902-1G OR943902-250MG OR943902-5G
APOSOR943902-1GEA 3350 SEK
APOSOR943902-1G APOSOR943902-250MG APOSOR943902-5G
5,6-Dimethylnicotinonitrile 97%
5,6-Dimethylnicotinonitrile
Formel: C₈H₈N₂
MW: 132,163 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11101052
CAS nummer: 113124-09-7