C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine 95%
Leverantör: Apollo Scientific
Synonyms: (7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)methanamine
OR943848-1G OR943848-250MG
APOSOR943848-1GEA 9500 SEK
APOSOR943848-1G APOSOR943848-250MG
C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine 95%
C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine
Formel: C₈H₁₀N₄
MW: 162,192 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09994896
CAS nummer: 1020033-70-8