Di(pyridin-3-yl)methanone 95%
Leverantör: Apollo Scientific
OR943214-1G OR943214-250MG
APOSOR943214-1GEA 3400 SEK
APOSOR943214-1G APOSOR943214-250MG
Di(pyridin-3-yl)methanone 95%
Di(pyridin-3-yl)methanone
Formel: C₁₁H₈N₂O
MW: 184,194 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01464076
CAS nummer: 35779-35-2