5,8-Dihydro-1-naphthol 87%
Leverantör: Apollo Scientific
OR943087-1G OR943087-250MG
APOSOR943087-1GEA 4275 SEK
APOSOR943087-1G APOSOR943087-250MG
5,8-Dihydro-1-naphthol 87%
5,8-Dihydro-1-naphthol
Formel: C₁₀H₁₀O
MW: 146.186 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00128247
CAS nummer: 27673-48-9