Ethyl 3-hydroxy-2,2-dimethylpropanoate 95%
Leverantör: Apollo Scientific
OR941920-1G OR941920-250MG
APOSOR941920-1GEA 755 SEK
APOSOR941920-1G APOSOR941920-250MG
Ethyl 3-hydroxy-2,2-dimethylpropanoate 95%
Ethyl 3-hydroxy-2,2-dimethylpropanoate
Formel: C₇H₁₄O₃
MW: 146,184 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD21643853
CAS nummer: 14002-73-4