3-Bromocinnolin-4-ol > 97%

Leverantör: Apollo Scientific

OR940970-1G OR940970-250MG
APOSOR940970-1GEA 3500 SEK
APOSOR940970-1G APOSOR940970-250MG
3-Bromocinnolin-4-ol > 97%
3-Bromocinnolin-4-ol
Formel: C₈H₅BrN₂O
Storage Temperature: Frys
MDL Number: MFCD20921550
CAS nummer: 19419-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR