Ethyl 2-((methoxycarbonyl)amino)acetate 95%

Leverantör: Apollo Scientific

OR939896-100MG OR939896-250MG OR939896-500MG
APOSOR939896-100MGEA 2250 SEK
APOSOR939896-100MG APOSOR939896-250MG APOSOR939896-500MG
Ethyl 2-((methoxycarbonyl)amino)acetate 95%
Ethyl 2-((methoxycarbonyl)amino)acetate
Formel: C₆H₁₁NO₄
MW: 161,156 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03384670
CAS nummer: 5602-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR