Allyl 1H-imidazole-1-carboxylate 95%
Leverantör: Apollo Scientific
OR939727-1G OR939727-25G OR939727-5G
APOSOR939727-1GEA 1200 SEK
APOSOR939727-1G APOSOR939727-25G APOSOR939727-5G
Allyl 1H-imidazole-1-carboxylate 95%
Allyl 1H-imidazole-1-carboxylate
Formel: C₇H₈N₂O₂
MW: 152.151 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18835431
CAS nummer: 83395-39-5