Cyclohexanone-4-carboxaldehyde 95%
Leverantör: Apollo Scientific
Synonyms: 4-Oxocyclohexanecarbaldehyde
OR939610-1G OR939610-250MG
APOSOR939610-1GEA 6250 SEK
APOSOR939610-1G APOSOR939610-250MG
Cyclohexanone-4-carboxaldehyde 95%
Cyclohexanone-4-carboxaldehyde
Formel: C₇H₁₀O₂
MW: 126,153 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04972553
CAS nummer: 96184-81-5