3-Methoxypropanoyl chloride 95%
Leverantör: Apollo Scientific
OR939440-1G OR939440-250MG OR939440-500MG
APOSOR939440-1GEA 1375 SEK
APOSOR939440-1G APOSOR939440-250MG APOSOR939440-500MG
3-Methoxypropanoyl chloride 95%
3-Methoxypropanoyl chloride
Formel: C₄H₇ClO₂
Storage Temperature: Omgivande
MDL Number: MFCD03424697
CAS nummer: 4244-59-1