Ethyl 2-methyl-4h-furo[3,2-b]pyrrole-5-carboxylate 95%
Leverantör: Apollo Scientific
OR939371-1G OR939371-250MG
APOSOR939371-1GEA 7875 SEK
APOSOR939371-1G APOSOR939371-250MG
Ethyl 2-methyl-4h-furo[3,2-b]pyrrole-5-carboxylate 95%
Ethyl 2-methyl-4h-furo[3,2-b]pyrrole-5-carboxylate
Formel: C₁₀H₁₁NO₃
Storage Temperature: Omgivande
MDL Number: MFCD00179150
CAS nummer: 80709-78-0