Boc-2-Cyclohexylglycine 97%

Leverantör: Apollo Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid

OR939277-1G OR939277-25G OR939277-5G
APOSOR939277-1GEA 670 SEK
APOSOR939277-1G APOSOR939277-25G APOSOR939277-5G
Boc-2-Cyclohexylglycine 97%
Boc-2-Cyclohexylglycine
Formel: C₁₃H₂₃NO₄
MW: 257,326 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04115511
CAS nummer: 35264-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR