6-Chlorooxazolo[4,5-b]pyridin-2(3H)-one 95%

Leverantör: Apollo Scientific

OR939132-1G OR939132-250MG
APOSOR939132-1GEA 685 SEK
APOSOR939132-1G APOSOR939132-250MG
6-Chlorooxazolo[4,5-b]pyridin-2(3H)-one 95%
6-Chlorooxazolo[4,5-b]pyridin-2(3H)-one
Formel: C₆H₃ClN₂O₂
MW: 170,553 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14706178
CAS nummer: 35570-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR