3-Methoxy-5-methylbenzaldehyde 95%

Leverantör: Apollo Scientific

OR938577-1G OR938577-250MG
APOSOR938577-1GEA 805 SEK
APOSOR938577-1G APOSOR938577-250MG
3-Methoxy-5-methylbenzaldehyde 95%
3-Methoxy-5-methylbenzaldehyde
Formel: C₉H₁₀O₂
MW: 150,175 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11046639
CAS nummer: 90674-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR