5-Bromo-1H-pyrazolo[4,3-b]pyridine 97%
Leverantör: Apollo Scientific
OR933142-1G OR933142-250MG
APOSOR933142-1GEA 1150 SEK
APOSOR933142-1G APOSOR933142-250MG
5-Bromo-1H-pyrazolo[4,3-b]pyridine 97%
5-Bromo-1H-pyrazolo[4,3-b]pyridine
Formel: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD15526714
CAS nummer: 1227628-78-5