Methyl 5-bromo-4-methoxypyridine-2-carboxylate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 5-bromo-4-methoxypicolinate
OR933037-1G OR933037-250MG OR933037-5G
APOSOR933037-1GEA 345 SEK
APOSOR933037-1G APOSOR933037-250MG APOSOR933037-5G
Methyl 5-bromo-4-methoxypyridine-2-carboxylate 97%
Methyl 5-bromo-4-methoxypyridine-2-carboxylate
Formel: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18257513
CAS nummer: 1256789-95-3