2-Phenylisoindolin-1-one 95%
Leverantör: Apollo Scientific
Varning
OR932601-1G OR932601-250MG
APOSOR932601-1GEA 5075 SEK
APOSOR932601-1G APOSOR932601-250MG
2-Phenylisoindolin-1-one 95%
2-Phenylisoindolin-1-one
Formel: C₁₄H₁₁NO
Storage Temperature: Omgivande
MDL Number: MFCD00023020
CAS nummer: 5388-42-1