5,6-Dichloropyridazin-3(2H)-one 95%
Leverantör: Apollo Scientific
OR932432-1G
OR932432-5G
APOSOR932432-1GEA
875
SEK
APOSOR932432-1G
APOSOR932432-5G
5,6-Dichloropyridazin-3(2H)-one 95%
5,6-Dichloropyridazin-3(2H)-one
Formel:
C₄H₂Cl₂N₂O MW: 164,977 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD04971420 CAS nummer: 17285-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...