(2-Chloro-phenyl)-oxo-acetonitrile 95%
Leverantör: Apollo Scientific
Synonyms: 2-Chlorobenzoyl cyanide
OR932078-1G OR932078-5G
APOSOR932078-1GEA 2050 SEK
APOSOR932078-1G APOSOR932078-5G
(2-Chloro-phenyl)-oxo-acetonitrile 95%
(2-Chloro-phenyl)-oxo-acetonitrile
Formel: C₈H₄ClNO
MW: 165.576 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03092702
CAS nummer: 35022-42-5