4H,5H,6H,7H,8H-Thieno[3,2-b]azepin-5-one 95%
Leverantör: Apollo Scientific
OR931890-1G OR931890-250MG
APOSOR931890-1GEA 1200 SEK
APOSOR931890-1G APOSOR931890-250MG
4H,5H,6H,7H,8H-Thieno[3,2-b]azepin-5-one 95%
4H,5H,6H,7H,8H-Thieno[3,2-b]azepin-5-one
Formel: C₈H₉NOS
MW: 167,228 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00980799
CAS nummer: 4751-61-5