(1-Methyl-2-oxoethyl)carbamic acid tert-butyl ester 95%
Leverantör: Apollo Scientific
Synonyms: tert-Butyl (1-oxopropan-2-yl)carbamate
OR931836-1G OR931836-250MG
APOSOR931836-1GEA 5725 SEK
APOSOR931836-1G APOSOR931836-250MG
(1-Methyl-2-oxoethyl)carbamic acid tert-butyl ester 95%
(1-Methyl-2-oxoethyl)carbamic acid tert-butyl ester
Formel: C₈H₁₅NO₃
MW: 173.21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12031700
CAS nummer: 114857-00-0