1H-Pyrrole-2,3-dicarboxylic acid dimethyl ester 95%
Leverantör: Apollo Scientific
Synonyms: Dimethyl 1H-pyrrole-2,3-dicarboxylate
OR931157-1G OR931157-250MG
APOSOR931157-1GEA 5675 SEK
APOSOR931157-1G APOSOR931157-250MG
1H-Pyrrole-2,3-dicarboxylic acid dimethyl ester 95%
1H-Pyrrole-2,3-dicarboxylic acid dimethyl ester
Formel: C₈H₉NO₄
MW: 183.161 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02328549
CAS nummer: 2818-08-8