2-(7-Methyl-1H-indol-3-yl)acetonitrile 95%
Leverantör: Apollo Scientific
OR931100-1G OR931100-250MG
APOSOR931100-1GEA 7625 SEK
APOSOR931100-1G APOSOR931100-250MG
2-(7-Methyl-1H-indol-3-yl)acetonitrile 95%
2-(7-Methyl-1H-indol-3-yl)acetonitrile
Formel: C₁₁H₁₀N₂
MW: 170,211 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04972089
CAS nummer: 858232-97-0