6-Bromo-1,2,4-triazin-3-amine 98%
Leverantör: Apollo Scientific
Synonyms:
3-Amino-6-bromo-1,2,4-triazine
OR930027-1G
OR930027-25G
OR930027-5G
APOSOR930027-1GEA
515
SEK
APOSOR930027-1G
APOSOR930027-25G
APOSOR930027-5G
6-Bromo-1,2,4-triazin-3-amine 98%
6-Bromo-1,2,4-triazin-3-amine
Formel:
C₃H₃BrN₄ MW: 174,987 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD16877209 CAS nummer: 69249-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...