N,N-Dimethyl-4-nitrobenzylamine 98%
Leverantör: Apollo Scientific
Synonyms: N,N-Dimethyl-1-(4-nitrophenyl)methanamine
OR929714-10G OR929714-1G OR929714-5G
APOSOR929714-10GEA 4875 SEK
APOSOR929714-10G APOSOR929714-1G APOSOR929714-5G
N,N-Dimethyl-4-nitrobenzylamine 98%
N,N-Dimethyl-4-nitrobenzylamine
Formel: C₉H₁₂N₂O₂
MW: 180,204 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00035956
CAS nummer: 15184-96-0