7-Methoxy-3,4-dihydro-2H-isoquinolin-1-one 95%
Leverantör: Apollo Scientific
Synonyms: 7-Methoxy-3,4-dihydroisoquinolin-1(2H)-one
OR929617-1G OR929617-250MG
APOSOR929617-1GEA 1900 SEK
APOSOR929617-1G APOSOR929617-250MG
7-Methoxy-3,4-dihydro-2H-isoquinolin-1-one 95%
7-Methoxy-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04114863
CAS nummer: 22246-04-4