2-Isopropyl-4-pyrimidinecarbaldehyde 95%

Leverantör: Apollo Scientific

OR929594-1G OR929594-250MG
APOSOR929594-1GEA 8475 SEK
APOSOR929594-1G APOSOR929594-250MG
2-Isopropyl-4-pyrimidinecarbaldehyde 95%
2-Isopropyl-4-pyrimidinecarbaldehyde
Formel: C₈H₁₀N₂O
MW: 150,178 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10696974
CAS nummer: 944901-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR