3-(Propan-2-yloxy)azetidine hydrochloride
Leverantör: Apollo Scientific
Synonyms: 3-Isopropoxyazetidine hydrochloride
OR928975-25G OR928975-50G OR928975-5G
APOSOR928975-25GEA 3650 SEK
APOSOR928975-25G APOSOR928975-50G APOSOR928975-5G
3-(Propan-2-yloxy)azetidine hydrochloride
3-(Propan-2-yloxy)azetidine hydrochloride
Formel: C₆H₁₃NO·ClH
MW: 151,634 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06804558
CAS nummer: 871657-49-7