1-Acetylcyclohexanol 95%

Leverantör: Apollo Scientific

OR928833-1G OR928833-250MG
APOSOR928833-1GEA 8475 SEK
APOSOR928833-1G APOSOR928833-250MG
1-Acetylcyclohexanol 95%
1-Acetylcyclohexanol
Formel: C₈H₁₄O₂
Storage Temperature: Omgivande
MDL Number: MFCD00019352
CAS nummer: 1123-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR