α-acetoxyphenylacetonitrile 97%
Leverantör: Apollo Scientific
OR928402-1G
OR928402-5G
APOSOR928402-1GEA
2575
SEK
APOSOR928402-1G
APOSOR928402-5G
α-acetoxyphenylacetonitrile 97%
α-acetoxyphenylacetonitrile
Formel:
C₁₀H₉NO₂ Storage Temperature: Omgivande |
MDL Number:
MFCD22205885 CAS nummer: 5762-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...