5-Benzothiazolecarboxaldehyde 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Benzo[d]thiazole-5-carbaldehyde
OR927910-100MG OR927910-1G OR927910-250MG
APOSOR927910-100MGEA 615 SEK
APOSOR927910-100MG APOSOR927910-1G APOSOR927910-250MG
5-Benzothiazolecarboxaldehyde 95%
5-Benzothiazolecarboxaldehyde
Formel: C₈H₅NOS
Storage Temperature: Omgivande
MDL Number: MFCD11858591
CAS nummer: 394223-38-2