6-Chloro-2-methylimidazo[1,2-b]pyridazine 95%
Leverantör: Apollo Scientific
OR927789-10G OR927789-1G OR927789-5G
APOSOR927789-10GEA 3525 SEK
APOSOR927789-10G APOSOR927789-1G APOSOR927789-5G
6-Chloro-2-methylimidazo[1,2-b]pyridazine 95%
6-Chloro-2-methylimidazo[1,2-b]pyridazine
Formel: C₇H₆ClN₃
MW: 167,596 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07778346
CAS nummer: 14793-00-1