5,8-Dibromoimidazo[1,2-a]pyrazine 97%
Leverantör: Apollo Scientific
Varning
OR926842-100MG OR926842-250MG
APOSOR926842-100MGEA 855 SEK
APOSOR926842-100MG APOSOR926842-250MG
5,8-Dibromoimidazo[1,2-a]pyrazine 97%
5,8-Dibromoimidazo[1,2-a]pyrazine
Formel: C₆H₃Br₂N₃
MW: 276,916 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD13193554
CAS nummer: 957344-74-0