2-Hydroxy-4-phenylthiazole 95%

Leverantör: Apollo Scientific

OR926235-1G OR926235-5G
APOSOR926235-1GEA 1200 SEK
APOSOR926235-1G APOSOR926235-5G
2-Hydroxy-4-phenylthiazole 95%
2-Hydroxy-4-phenylthiazole
Formel: C₉H₇NOS
MW: 177,223 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00665954
CAS nummer: 3884-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR